Thermo Scientific Chemicals

4-Chlorobenzophenone, 99%, Thermo Scientific Chemicals

Catalog number: 108691000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

4-Chlorobenzophenone, 99%, Thermo Scientific Chemicals

Catalog number: 108691000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 108691000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
134-85-0
IUPAC Name
(4-chlorophenyl)(phenyl)methanone
Molecular Formula
C13H9ClO
InChI Key
UGVRJVHOJNYEHR-UHFFFAOYSA-N
SMILES
ClC1=CC=C(C=C1)C(=O)C1=CC=CC=C1
Infrared spectrum
Conforms
Appearance (Color)
White to off-white
GC
>=98.5 %
Appearance (Form)
Powder
Melting point
75°C to 77°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text