4-Chlorobenzotrifluoride, 98%, Thermo Scientific Chemicals
4-Chlorobenzotrifluoride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Chlorobenzotrifluoride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Catalog number 108732500
Price (USD)/ Each
-
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name1-chloro-4-(trifluoromethyl)benzene
Molecular FormulaC7H4ClF3
InChI KeyQULYNCCPRWKEMF-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC=C(Cl)C=C1
Molecular Weight (g/mol)180.55
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4453 to 1.4473 (20°C, 589 nm)
Appearance (Color)Clear colorless
Infrared spectrumConforms
GC>=97.5 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only