Thermo Scientific Chemicals

3-Chlorobenzoyl chloride, 99+%, Thermo Scientific Chemicals

Catalog number: 108741000
100 mL, Glass bottle, Each
Thermo Scientific Chemicals

3-Chlorobenzoyl chloride, 99+%, Thermo Scientific Chemicals

Catalog number: 108741000
100 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 108741000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
618-46-2
IUPAC Name
3-chlorobenzoyl chloride
Molecular Formula
C7H4Cl2O
InChI Key
WHIHIKVIWVIIER-UHFFFAOYSA-N
SMILES
ClC(=O)C1=CC=CC(Cl)=C1
Appearance (Color)
Clear colorless
Infrared spectrum
Conforms
Titration RCOOH
=<1.0 % (as Chlorobenzoic acid)
GC
=<0.03 % Total other poly-halogenated acid halides
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text