4-Chlorobenzyl alcohol, 99%, Thermo Scientific Chemicals
4-Chlorobenzyl alcohol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Chlorobenzyl alcohol, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 108770250
Price (USD)/ Each
64.20
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
64.20
Add to cart
4-Chlorobenzyl alcohol, 99%, Thermo Scientific Chemicals
Catalog number108770250
Price (USD)/ Each
64.20
-
Add to cart
Chemical Identifiers
CAS873-76-7
IUPAC Name(4-chlorophenyl)methanol
Molecular FormulaC7H7ClO
InChI KeyPTHGDVCPCZKZKR-UHFFFAOYSA-N
SMILESOCC1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or needles
Melting point69°C to 72°C
Appearance (Color)White to beige
Infrared spectrumConforms
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only