4-Chlorobenzyl cyanide, 98+%, Thermo Scientific Chemicals
4-Chlorobenzyl cyanide, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Chlorobenzyl cyanide, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
Packaging:
Glass bottle
Catalog number 108811000
Price (USD)/ Each
28.70
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
28.70
Add to cart
4-Chlorobenzyl cyanide, 98+%, Thermo Scientific Chemicals
Catalog number108811000
Price (USD)/ Each
28.70
-
Add to cart
Chemical Identifiers
CAS140-53-4
IUPAC Name2-(4-chlorophenyl)acetonitrile
Molecular FormulaC8H6ClN
InChI KeyIVYMIRMKXZAHRV-UHFFFAOYSA-N
SMILESClC1=CC=C(CC#N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid after melting
Infrared spectrumConforms
GC>=98.0 %
Water=<0.3 % (K.F.)
Refractive index1.5415 to 1.5435 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only