2-Chlorobutane, 99+%
2-Chlorobutane, 99+%
2-Chlorobutane, 99+%
2-Chlorobutane, 99+%
Thermo Scientific Chemicals

2-Chlorobutane, 99+%

CAS: 78-86-4 | C4H9Cl | 92.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Catalog number 108832500
Price (USD)
41.65
Special offer
Online exclusive
Ends: 26-Sep-2025
49.10
Save 7.45 (15%)
Each
In stock
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
41.65
Special offer
Online exclusive
Ends: 26-Sep-2025
49.10
Save 7.45 (15%)
Each
Add to cart
Chemical Identifiers
CAS78-86-4
IUPAC Name2-chlorobutane
Molecular FormulaC4H9Cl
InChI KeyBSPCSKHALVHRSR-UHFFFAOYNA-N
SMILESCCC(C)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Refractive index1.3960 to 1.3980 (20°C, 589 nm)
Infrared spectrumConforms
GC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection