Thermo Scientific Chemicals

4-Chlorobutyryl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 108900010
1 L, Each
Thermo Scientific Chemicals

4-Chlorobutyryl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 108900010
1 L, Each
Quantity
Catalog number: 108900010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
4635-59-0
IUPAC Name
4-chlorobutanoyl chloride
Molecular Formula
C4H6Cl2O
InChI Key
CDIIZULDSLKBKV-UHFFFAOYSA-N
SMILES
ClCCCC(Cl)=O
Appearance (Color)
Clear colorless to yellow
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=97.5 %
Refractive index
1.4605 to 1.4625 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text