Thermo Scientific Chemicals

4-Chlorocinnamic acid, 99%, predominantly trans, Thermo Scientific Chemicals

Catalog number: 108941000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

4-Chlorocinnamic acid, 99%, predominantly trans, Thermo Scientific Chemicals

Catalog number: 108941000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 108941000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1615-02-7
Appearance (Color)
White
Melting point
246°C to 250°C
Titration with NaOH
98.5 to 101.5 %
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text