Thermo Scientific Chemicals

4-Chloro-3,5-dimethylphenol, 99%, Thermo Scientific Chemicals

Catalog number: 109061000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

4-Chloro-3,5-dimethylphenol, 99%, Thermo Scientific Chemicals

Catalog number: 109061000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 109061000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
88-04-0
IUPAC Name
4-chloro-3,5-dimethylphenol
Molecular Formula
C8H9ClO
InChI Key
OSDLLIBGSJNGJE-UHFFFAOYSA-N
SMILES
CC1=CC(O)=CC(C)=C1Cl
Appearance (Color)
White to light beige
Appearance (Form)
Crystalline powder or crystals
Infrared spectrum
Conforms
Melting point
113°C to 116°C
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text