Thermo Scientific Chemicals

Chlorodiphenylphosphine, 95%, tech., Thermo Scientific Chemicals

Catalog number: 109130250
25 mL, Glass bottle, Each
Thermo Scientific Chemicals

Chlorodiphenylphosphine, 95%, tech., Thermo Scientific Chemicals

Catalog number: 109130250
25 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 109130250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1079-66-9
IUPAC Name
chlorodiphenylphosphane
Molecular Formula
C12H10ClP
InChI Key
XGRJZXREYAXTGV-UHFFFAOYSA-N
SMILES
ClP(C1=CC=CC=C1)C1=CC=CC=C1
Appearance
May develop some turbidity or precipitate
Infrared spectrum
Conforms
GC
>=94 %
Refractive index
1.6340 to 1.6370 (20°C, 589 nm)
Appearance (Color)
Yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text