Thermo Scientific Chemicals

N-(2-Chloroethyl)morpholine hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: 109200050
5 g, Each
Thermo Scientific Chemicals

N-(2-Chloroethyl)morpholine hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: 109200050
5 g, Each
Quantity
Catalog number: 109200050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
3647-69-6
IUPAC Name
hydrogen 4-(2-chloroethyl)morpholine chloride
Molecular Formula
C6H13Cl2NO
InChI Key
NBJHDLKSWUDGJG-UHFFFAOYSA-N
SMILES
[H+].[Cl-].ClCCN1CCOCC1
Infrared spectrum
Conforms
Melting point
182°C to 185°C (on dry substance)
Titration with HClO4
>=98.5 % (On dry substance)
Loss on drying
=<1 % (80°C)
Appearance (Color)
White to beige

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text