Thermo Scientific Chemicals

1-(2-Chloroethyl)piperidine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 109211000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

1-(2-Chloroethyl)piperidine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 109211000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 109211000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2008-75-5
IUPAC Name
hydrogen 1-(2-chloroethyl)piperidine chloride
Molecular Formula
C7H15Cl2N
InChI Key
VFLQQZCRHPIGJU-UHFFFAOYSA-N
SMILES
[H+].[Cl-].ClCCN1CCCCC1
Appearance (Color)
White to yellow to beige
Appearance (Form)
Powder or crystals
Infrared spectrum
Conforms
Melting point
228°C to 234°C
Titration with HClO4
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text