N-(2-Chloroethyl)pyrrolidine hydrochloride, 98%, Thermo Scientific Chemicals
N-(2-Chloroethyl)pyrrolidine hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-(2-Chloroethyl)pyrrolidine hydrochloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 109220250
Price (USD)/ Each
52.40
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
52.40
Add to cart
N-(2-Chloroethyl)pyrrolidine hydrochloride, 98%, Thermo Scientific Chemicals
Catalog number109220250
Price (USD)/ Each
52.40
-
Add to cart
Chemical Identifiers
CAS7250-67-1
IUPAC Namehydrogen 1-(2-chloroethyl)pyrrolidine chloride
Molecular FormulaC6H13Cl2N
InChI KeyFSNGFFWICFYWQC-UHFFFAOYSA-N
SMILES[H+].[Cl-].ClCCN1CCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Titration with HClO4>=97.5 %
Melting point165°C to 175°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only