Thermo Scientific Chemicals

5-Chloro-2-methylaniline, 98%, Thermo Scientific Chemicals

Catalog number: 109385000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

5-Chloro-2-methylaniline, 98%, Thermo Scientific Chemicals

Catalog number: 109385000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 109385000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
95-79-4
IUPAC Name
5-chloro-2-methylaniline
Molecular Formula
C7H8ClN
InChI Key
WRZOMWDJOLIVQP-UHFFFAOYSA-N
SMILES
CC1=CC=C(Cl)C=C1N
Appearance
May darken on storage
GC
>=97.5 %
Appearance (Color)
Clear yellow to light brown
Refractive index
1.5830 to 1.5850 (20°C, 589 nm)
Appearance (Form)
Liquid after melting

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text