1-(Chloromethyl)naphthalene, 95%, Thermo Scientific Chemicals
1-(Chloromethyl)naphthalene, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-(Chloromethyl)naphthalene, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 109450050
Price (USD)/ Each
21.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
25.00 
Save 3.35 (13%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
21.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
25.00 
Save 3.35 (13%)
Add to cart
1-(Chloromethyl)naphthalene, 95%, Thermo Scientific Chemicals
Catalog number109450050
Price (USD)/ Each
21.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
25.00 
Save 3.35 (13%)
-
Add to cart
Chemical Identifiers
IUPAC Name1-(chloromethyl)naphthalene
Molecular FormulaC11H9Cl
InChI KeyXMWGTKZEDLCVIG-UHFFFAOYSA-N
SMILESClCC1=C2C=CC=CC2=CC=C1
Molecular Weight (g/mol)176.64
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid after melting
HPLCRemainder 2-chloromethylnaphtalene
Infrared spectrumConforms
Appearance (Color)Clear yellow to brown
HPLC>=94 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only