1-(Chloromethyl)naphthalene, 95%, Thermo Scientific Chemicals
1-(Chloromethyl)naphthalene, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-(Chloromethyl)naphthalene, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
Packaging:
Glass bottle
Catalog number 109455000
Price (USD)/ Each
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name1-(chloromethyl)naphthalene
Molecular FormulaC11H9Cl
InChI KeyXMWGTKZEDLCVIG-UHFFFAOYSA-N
SMILESClCC1=C2C=CC=CC2=CC=C1
Molecular Weight (g/mol)176.64
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid after melting
HPLCRemainder 2-chloromethylnaphtalene
Infrared spectrumConforms
Appearance (Color)Clear yellow to brown
HPLC>=94 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only