Thermo Scientific Chemicals

4-Chloro-3-methylphenol, 99+%, Thermo Scientific Chemicals

Catalog number: 109485000
500 g, Each
Thermo Scientific Chemicals

4-Chloro-3-methylphenol, 99+%, Thermo Scientific Chemicals

Catalog number: 109485000
500 g, Each
Quantity
Catalog number: 109485000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
59-50-7
IUPAC Name
4-chloro-3-methylphenol
Molecular Formula
C7H7ClO
InChI Key
CFKMVGJGLGKFKI-UHFFFAOYSA-N
SMILES
CC1=CC(O)=CC=C1Cl
Appearance (Color)
White to almost white
Melting point
64°C to 67°C
Infrared spectrum
Conforms
GC
>=99.0 %
Appearance (Form)
Crystals, flakes, pellets or crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text