Thermo Scientific Chemicals

2-(Chloromethyl)quinoline hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: 109510250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

2-(Chloromethyl)quinoline hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: 109510250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 109510250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3747-74-8
IUPAC Name
hydrogen 2-(chloromethyl)quinoline chloride
Molecular Formula
C10H9Cl2N
InChI Key
WDETYCRYUBGKCE-UHFFFAOYSA-N
SMILES
[H+].[Cl-].ClCC1=CC=C2C=CC=CC2=N1
Infrared spectrum
Conforms
Titration Argentometric
>=96.0 %
Appearance (Color)
Cream to light yellow
Titration with HClO4
>=96.0 %
Solubility
(5 % in water) Clear

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text