1-Chloro-4-nitrobenzene, 99%
1-Chloro-4-nitrobenzene, 99%
1-Chloro-4-nitrobenzene, 99%
1-Chloro-4-nitrobenzene, 99%
Thermo Scientific Chemicals

1-Chloro-4-nitrobenzene, 99%

CAS: 100-00-5 | C6H4ClNO2 | 157.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 g
100 g
Packaging:
View all
Plastic bottle
Catalog number 109631000
Price (USD)
31.90
Each
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
31.90
Each
Add to cart
Chemical Identifiers
CAS100-00-5
IUPAC Name1-chloro-4-nitrobenzene
Molecular FormulaC6H4ClNO2
InChI KeyCZGCEKJOLUNIFY-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals or flakes
Appearance (Color)Yellow
Infrared spectrumConforms
GC>=98.5 %
Melting point81°C to 84°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Share catalog number, name or link