2-Chloro-4-nitrobenzoic acid, 98%, Thermo Scientific Chemicals
2-Chloro-4-nitrobenzoic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Chloro-4-nitrobenzoic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 109651000
Price (USD)/ Each
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
IUPAC Name2-chloro-4-nitrobenzoic acid
Molecular FormulaC7H4ClNO4
InChI KeyQAYNSPOKTRVZRC-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(C=C1Cl)[N+]([O-])=O
Molecular Weight (g/mol)201.56
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals or needles
Melting point138°C to 144°C
Infrared spectrumConforms
Appearance (Color)Yellow
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only