Thermo Scientific Chemicals

2-Chloro-3-nitropyridine, 99+%, Thermo Scientific Chemicals

Catalog number: 109731000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Chloro-3-nitropyridine, 99+%, Thermo Scientific Chemicals

Catalog number: 109731000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 109731000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5470-18-8
IUPAC Name
2-chloro-3-nitropyridine
Molecular Formula
C5H3ClN2O2
InChI Key
UUOLETYDNTVQDY-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC=CN=C1Cl
Appearance (Color)
Yellow
Appearance (Form)
Crystalline powder
Melting point
100°C to 103°C
Infrared spectrum
Conforms
HPLC
>=99.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text