Informational:Inquire about OEM or Commercial Supply version of this product here.
Invitrogen™

UltraPure™ DNase/RNase-Free Distilled Water

Catalog number: 10977035
Have Questions?
Invitrogen™

UltraPure™ DNase/RNase-Free Distilled Water

Catalog number: 10977035
Have Questions?
Catalog Number
10977035
Unit Size
500 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
10977035500 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
UltraPure™ DNase/RNase-Free Distilled Water is designed for use in all molecular biology applications. It is 0.1-μm membrane-filtered and tested for DNase and RNase activity.

Performance and quality testing
No DNase, RNase, or protease activity detected. Our distilled water system is routinely monitored for compliance with current USP monograph test requirements for Water for Injection (WFI).
For Research Use Only. Not for use in diagnostic procedures.

Specifications

pH
6 to 8
Recommended Storage
May be stored at room temperature.
Grade
Molecular Biology
Packaging
Bottle
Treatment(s)
Not DEPC-Treated
Quantity
500 mL
Purification Method
Membrane-Filtered
Shipping Condition
Room Temperature
Product Line
UltraPure™
Green Features
Sustainable packaging

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text