3-chlorophenylacetic acid, 99+%, Thermo Scientific Chemicals
3-chlorophenylacetic acid, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-chlorophenylacetic acid, 99+%, Thermo Scientific Chemicals

Have Questions?
Quantity:
10 g
Packaging:
Glass bottle
Catalog number 109840100
Price (USD)/ Each
80.60
-
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
80.60
Add to cart
3-chlorophenylacetic acid, 99+%, Thermo Scientific Chemicals
Catalog number109840100
Price (USD)/ Each
80.60
-
Add to cart
Chemical Identifiers
CAS1878-65-5
IUPAC Name2-(3-chlorophenyl)acetic acid
Molecular FormulaC8H7ClO2
InChI KeyWFPMUFXQDKMVCO-UHFFFAOYSA-N
SMILESOC(=O)CC1=CC=CC(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Glistening flakes and/or chunks
Titration with NaOH>=99.0 %
Appearance (Color)White
Infrared spectrumConforms
Melting point77°C to 80°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only