Thermo Scientific Chemicals

2-(4-Chlorophenyl)ethylamine, 98%, Thermo Scientific Chemicals

Catalog number: 109880050
5 mL, Glass bottle, Each
Thermo Scientific Chemicals

2-(4-Chlorophenyl)ethylamine, 98%, Thermo Scientific Chemicals

Catalog number: 109880050
5 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 109880050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
156-41-2
IUPAC Name
2-(4-chlorophenyl)ethan-1-aminium
Molecular Formula
C8H11ClN
InChI Key
SRXFXCKTIGELTI-UHFFFAOYSA-O
SMILES
[NH3+]CCC1=CC=C(Cl)C=C1
Infrared spectrum
Conforms
Water
=<0.5 % (K.F.)
Appearance (Color)
Clear colorless to yellow
Appearance (Form)
Liquid
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text