Thermo Scientific Chemicals

4-(4-Chlorophenyl)-1,2,3,6-tetrahydropyridine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: 109930050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

4-(4-Chlorophenyl)-1,2,3,6-tetrahydropyridine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: 109930050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 109930050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
51304-61-1
IUPAC Name
hydrogen 4-(4-chlorophenyl)-1,2,3,6-tetrahydropyridine chloride
Molecular Formula
C11H13Cl2N
InChI Key
NGDJUNABWNTULX-UHFFFAOYSA-N
SMILES
[H+].[Cl-].ClC1=CC=C(C=C1)C1=CCNCC1
Infrared spectrum
Conforms
Titration with HClO4
>=96.0 %
Appearance (Color)
White to light yellow
Melting point
199°C to 204°C
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text