Thermo Scientific Chemicals

2-Chloropyridine, 99%, Thermo Scientific Chemicals

Catalog number: 110011000
100 mL, Glass bottle, Each
Thermo Scientific Chemicals

2-Chloropyridine, 99%, Thermo Scientific Chemicals

Catalog number: 110011000
100 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 110011000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
109-09-1
IUPAC Name
2-chloropyridine
Molecular Formula
C5H4ClN
InChI Key
OKDGRDCXVWSXDC-UHFFFAOYSA-N
SMILES
ClC1=CC=CC=N1
Appearance (Color)
Clear colorless to light yellow
GC
>=98.5 %
Refractive index
1.5310 to 1.5330 (20°C, 589 nm)
Infrared spectrum
Conforms
Water
=<0.2 % (K.F.)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text