1x1 image pixel for data collection
Choline Chloride, 99%
Choline Chloride, 99%
Choline Chloride, 99%
Choline Chloride, 99%
Thermo Scientific Chemicals

Choline Chloride, 99%

CAS: 67-48-1 | C5H14ClNO | 139.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
50 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 110290500
Price (USD)
26.65
Special offer
Online exclusive
Ends: 26-Sep-2025
33.20
Save 6.55 (20%)
Each
In stock
Add to cart
Quantity:
50 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
26.65
Special offer
Online exclusive
Ends: 26-Sep-2025
33.20
Save 6.55 (20%)
Each
Add to cart
Chemical Identifiers
CAS67-48-1
IUPAC Name(2-hydroxyethyl)trimethylazanium chloride
Molecular FormulaC5H14ClNO
InChI KeySGMZJAMFUVOLNK-UHFFFAOYSA-M
SMILES[Cl-].C[N+](C)(C)CCO
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<2 %
Appearance (Color)White
Infrared spectrumConforms
Appearance (Form)Adhering crystals
Titration Argentometric98.5 to 101.5 % (on dry substance)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection