Thermo Scientific Chemicals

Chrysin, 99+%, Thermo Scientific Chemicals

Catalog number: 110320050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Chrysin, 99+%, Thermo Scientific Chemicals

Catalog number: 110320050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 110320050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
480-40-0
IUPAC Name
5,7-dihydroxy-2-phenyl-4H-chromen-4-one
Molecular Formula
C15H10O4
InChI Key
RTIXKCRFFJGDFG-UHFFFAOYSA-N
SMILES
OC1=CC(O)=C2C(=O)C=C(OC2=C1)C1=CC=CC=C1
Infrared spectrum
Conforms
Loss on drying
=<1.0 %
Melting point
284°C to 290°C
Appearance (Color)
Yellow
HPLC
>=99.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text