Thermo Scientific Chemicals

1,8-Cineole, 99%, Thermo Scientific Chemicals

Catalog number: 110341000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

1,8-Cineole, 99%, Thermo Scientific Chemicals

Catalog number: 110341000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 110341000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
470-82-6
IUPAC Name
1,3,3-trimethyl-2-oxabicyclo[2.2.2]octane
Molecular Formula
C10H18O
InChI Key
WEEGYLXZBRQIMU-UHFFFAOYSA-N
SMILES
CC12CCC(CC1)C(C)(C)O2
Appearance (Form)
Liquid
GC
>=98.5 %
Solubility
(in ethanol (70% v/v)) 1:2.5
Appearance (Color)
Clear colorless to light yellow
Refractive index
1.4550 to 1.4700 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text