Thermo Scientific Chemicals

Cinnamamide, 97%, predominantly trans, Thermo Scientific Chemicals

Catalog number: 110360050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Cinnamamide, 97%, predominantly trans, Thermo Scientific Chemicals

Catalog number: 110360050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 110360050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
621-79-4
IUPAC Name
(2E)-3-phenylprop-2-enamide
Molecular Formula
C9H9NO
InChI Key
APEJMQOBVMLION-VOTSOKGWSA-N
SMILES
NC(=O)\C=C\C1=CC=CC=C1
Appearance (Form)
Crystalline powder or crystals
HPLC
>=96.0 %
Melting point
146°C to 150°C
Appearance (Color)
White to beige
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text