Cinnamoyl chloride, 98%, predominantly trans, Thermo Scientific Chemicals
Cinnamoyl chloride, 98%, predominantly trans, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cinnamoyl chloride, 98%, predominantly trans, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number 110395000
Price (USD)/ Each
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name(2E)-3-phenylprop-2-enoyl chloride
Molecular FormulaC9H7ClO
InChI KeyWOGITNXCNOTRLK-VOTSOKGWSA-N
SMILESClC(=O)\C=C\C1=CC=CC=C1
Molecular Weight (g/mol)166.60
View more
SpecificationsSpecification SheetSpecification Sheet
Titration RCOCl97.5 to 102.5 %
Appearance (Color)White to yellow to orange
Infrared spectrumConforms
Appearance (Form)Crystals and/or chunks or fused solid
Melting point33°C to 37°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only