Citraconic acid, 99+%
Citraconic acid, 99+%
Citraconic acid, 99+%
Citraconic acid, 99+%
Thermo Scientific Chemicals

Citraconic acid, 99+%

CAS: 498-23-7 | C5H6O4 | 130.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
View all
Glass bottle
Catalog number 110430050
Price (USD)
24.65
Online Exclusive
27.40
Save 2.75 (10%)
Each
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
24.65
Online Exclusive
27.40
Save 2.75 (10%)
Each
Add to cart
Chemical Identifiers
CAS498-23-7
IUPAC Name(2Z)-2-methylbut-2-enedioic acid
Molecular FormulaC5H6O4
InChI KeyHNEGQIOMVPPMNR-IHWYPQMZSA-N
SMILESC\C(=C\C(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Titration with NaOH>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection