Thermo Scientific Chemicals

Citral, 95%, mixture of cis and trans, Thermo Scientific Chemicals

Catalog number: 110441000
100 mL, Glass bottle, Each
Thermo Scientific Chemicals

Citral, 95%, mixture of cis and trans, Thermo Scientific Chemicals

Catalog number: 110441000
100 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 110441000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5392-40-5
IUPAC Name
(2E)-3,7-dimethylocta-2,6-dienal
Molecular Formula
C10H16O
InChI Key
WTEVQBCEXWBHNA-JXMROGBWSA-N
SMILES
CC(C)=CCC\C(C)=C\C=O
Appearance (Color)
Clear yellow
Density
(20/4°C) 0.886 to 0.893 kg/l
Infrared spectrum
Conforms
Refractive index
1.4860 to 1.4900 (20°C, 589 nm)
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text