Citronellol, 95%
Citronellol, 95%
Citronellol, 95%
Thermo Scientific Chemicals

Citronellol, 95%

CAS: 106-22-9 | C10H20O | 156.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
5 mL
500 mL
Packaging:
Glass bottle
Catalog number 110460050
Price (USD)
31.65
Online Exclusive
34.70
Save 3.05 (9%)
Each
-
Add to cart
Quantity:
5 mL
Packaging:
Glass bottle
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
31.65
Online Exclusive
34.70
Save 3.05 (9%)
Each
Add to cart
Chemical Identifiers
CAS106-22-9
IUPAC Name(3R)-3,7-dimethyloct-6-en-1-ol
Molecular FormulaC10H20O
InChI KeyQMVPMAAFGQKVCJ-SNVBAGLBSA-N
SMILESC[C@@H](CCO)CCC=C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=94 %
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Refractive index1.4540 to 1.4580 (20°C, 589 nm)
Infrared spectrumConforms
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0467209Certificate of AnalysisOct 29, 2024110460050, 110465000, 110461000
A0435703Certificate of AnalysisApr 25, 2024110460050, 110465000, 110461000
A0395034Certificate of AnalysisDec 15, 2022110460050, 110465000, 110461000
A0394485Certificate of AnalysisDec 15, 2022110460050, 110465000, 110461000
A0397852Certificate of AnalysisDec 15, 2022110460050, 110465000, 110461000
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection