Coumarin, 99+%
Coumarin, 99+%
Coumarin, 99+%
Thermo Scientific Chemicals

Coumarin, 99+%

CAS: 91-64-5 | C9H6O2 | 146.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 g
Packaging:
View all
Glass bottle
Catalog number 110530050
Price (USD)
27.65
Online Exclusive
30.20
Save 2.55 (8%)
Each
In stock
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
27.65
Online Exclusive
30.20
Save 2.55 (8%)
Each
Add to cart
Chemical Identifiers
CAS91-64-5
IUPAC Name2H-chromen-2-one
Molecular FormulaC9H6O2
InChI KeyZYGHJZDHTFUPRJ-UHFFFAOYSA-N
SMILESO=C1OC2=CC=CC=C2C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point68°C to 71°C
Appearance (Color)White
Infrared spectrumConforms
GC>=99.0 %
Appearance (Form)Crystalline powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection