Thermo Scientific Chemicals

Coumarin, 99+%, Thermo Scientific Chemicals

Catalog number: 110532500
250 g, Plastic bottle, Each
Thermo Scientific Chemicals

Coumarin, 99+%, Thermo Scientific Chemicals

Catalog number: 110532500
250 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 110532500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
91-64-5
IUPAC Name
2H-chromen-2-one
Molecular Formula
C9H6O2
InChI Key
ZYGHJZDHTFUPRJ-UHFFFAOYSA-N
SMILES
O=C1OC2=CC=CC=C2C=C1
Appearance (Color)
White
Infrared spectrum
Conforms
GC
>=99.0 %
Melting point
68°C to 71°C
Appearance (Form)
Crystalline powder or crystals

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text