Thermo Scientific Chemicals

m-Cresol, 99%, Thermo Scientific Chemicals

Catalog number: 110580100
10 kg, Metal Drum, Each
Thermo Scientific Chemicals

m-Cresol, 99%, Thermo Scientific Chemicals

Catalog number: 110580100
10 kg, Metal Drum, Each
Quantity
Packaging
Catalog number: 110580100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
108-39-4
IUPAC Name
3-methylphenol
Molecular Formula
C7H8O
InChI Key
RLSSMJSEOOYNOY-UHFFFAOYSA-N
SMILES
CC1=CC=CC(O)=C1
Appearance
May darken on storage
GC
>=98.5 %
Appearance (Color)
Clear colorless to light red
Infrared spectrum
Conforms
Appearance (Form)
Viscous liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text