m-Cresol, 99%, Thermo Scientific Chemicals
m-Cresol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

m-Cresol, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
1 kg
2.5 kg
10 kg
Packaging:
View all
Glass bottle
Catalog number 110585000
Price (USD)/ Each
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS108-39-4
IUPAC Name3-methylphenol
Molecular FormulaC7H8O
InChI KeyRLSSMJSEOOYNOY-UHFFFAOYSA-N
SMILESCC1=CC=CC(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
AppearanceMay darken on storage
GC>=98.5 %
Appearance (Color)Clear colorless to light red
Infrared spectrumConforms
Appearance (Form)Viscous liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only