Thermo Scientific Chemicals

Cumene, 98%, pure, Thermo Scientific Chemicals

Catalog number: 110630025
2.5 L, Glass bottle, Each
Thermo Scientific Chemicals

Cumene, 98%, pure, Thermo Scientific Chemicals

Catalog number: 110630025
2.5 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 110630025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
98-82-8
IUPAC Name
(propan-2-yl)benzene
Molecular Formula
C9H12
InChI Key
RWGFKTVRMDUZSP-UHFFFAOYSA-N
SMILES
CC(C)C1=CC=CC=C1
Appearance (Color)
Clear colorless
Refractive index
1.4890 to 1.4930 (20°C, 589 nm)
Appearance (Form)
Liquid
GC
>=97.5 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text