Copper(II) acetylacetonate, 98%, Thermo Scientific Chemicals
Copper(II) acetylacetonate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Copper(II) acetylacetonate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 110650250
Price (USD)/ Each
38.65
Online exclusive
43.10 
Save 4.45 (10%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
38.65
Online exclusive
43.10 
Save 4.45 (10%)
Add to cart
Copper(II) acetylacetonate, 98%, Thermo Scientific Chemicals
Catalog number110650250
Price (USD)/ Each
38.65
Online exclusive
43.10 
Save 4.45 (10%)
-
Add to cart
Chemical Identifiers
CAS13395-16-9
IUPAC Namecopper(2+) bis((2Z)-4-oxopent-2-en-2-olate)
Molecular FormulaC10H14CuO4
InChI KeyQYJPSWYYEKYVEJ-FDGPNNRMSA-L
SMILES[Cu++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Appearance (Color)Blue to blue-grey
Titration with Na2S2O397.5 to 102.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only