Thermo Scientific Chemicals

Cyanogen bromide, 97%, Thermo Scientific Chemicals

Catalog number: 110780250
25 g, Glass Bottle in Tin Can, Each
Thermo Scientific Chemicals

Cyanogen bromide, 97%, Thermo Scientific Chemicals

Catalog number: 110780250
25 g, Glass Bottle in Tin Can, Each
Quantity
Packaging
Catalog number: 110780250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
506-68-3
IUPAC Name
carbononitridic bromide
Molecular Formula
CBrN
InChI Key
ATDGTVJJHBUTRL-UHFFFAOYSA-N
SMILES
BrC#N
Appearance (Color)
White
Identification
Bromides : positive
Appearance (Form)
Crystals
Titration Argentometric
96.0 to 103.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text