Thermo Scientific Chemicals

4-Cyanopyridine, 98%, Thermo Scientific Chemicals

Catalog number: 110870025
2.5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

4-Cyanopyridine, 98%, Thermo Scientific Chemicals

Catalog number: 110870025
2.5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 110870025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
100-48-1
IUPAC Name
pyridine-4-carbonitrile
Molecular Formula
C6H4N2
InChI Key
GPHQHTOMRSGBNZ-UHFFFAOYSA-N
SMILES
N#CC1=CC=NC=C1
Appearance (Color)
White to yellow to beige to light brown
Appearance (Form)
Powder or crystals or flakes
Melting point
76°C to 81°C
Infrared spectrum
Conforms
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text