4-Cyanopyridine, 98%, Thermo Scientific Chemicals
4-Cyanopyridine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Cyanopyridine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number 110875000
Price (USD)/ Each
-
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS100-48-1
IUPAC Namepyridine-4-carbonitrile
Molecular FormulaC6H4N2
InChI KeyGPHQHTOMRSGBNZ-UHFFFAOYSA-N
SMILESN#CC1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to beige to light brown
Appearance (Form)Powder or crystals or flakes
Melting point76°C to 81°C
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only