Thermo Scientific Chemicals

Cyclobutanecarboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: 110900250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Cyclobutanecarboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: 110900250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 110900250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
3721-95-7
IUPAC Name
cyclobutanecarboxylic acid
Molecular Formula
C5H8O2
InChI Key
TXWOGHSRPAYOML-UHFFFAOYSA-N
SMILES
OC(=O)C1CCC1
Appearance (Form)
Liquid
Refractive index
1.4410 to 1.4450 (20°C, 589 nm)
Water
=<0.5 % (K.F.)
Appearance (Color)
Clear colorless to yellow
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text