Thermo Scientific Chemicals

Cyclobutanecarboxylic acid chloride, 99%, Thermo Scientific Chemicals

Catalog number: 110910500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

Cyclobutanecarboxylic acid chloride, 99%, Thermo Scientific Chemicals

Catalog number: 110910500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 110910500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
5006-22-4
IUPAC Name
cyclobutanecarbonyl chloride
Molecular Formula
C5H7ClO
InChI Key
JFWMYCVMQSLLOO-UHFFFAOYSA-N
SMILES
ClC(=O)C1CCC1
Appearance (Color)
Clear colorless to yellow
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Titration RCOCl
98.5 to 101.5 %
Refractive index
1.4540 to 1.4560 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text