Thermo Scientific Chemicals

1,1-Cyclobutanedicarboxylic acid, 99%, Thermo Scientific Chemicals

Catalog number: 110920050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

1,1-Cyclobutanedicarboxylic acid, 99%, Thermo Scientific Chemicals

Catalog number: 110920050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 110920050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
5445-51-2
IUPAC Name
cyclobutane-1,1-dicarboxylic acid
Molecular Formula
C6H8O4
InChI Key
CCQPAEQGAVNNIA-UHFFFAOYSA-N
SMILES
OC(=O)C1(CCC1)C(O)=O
Infrared spectrum
Conforms
Water
=<0.1 % (K.F.)
Clarity of solution
(5 w/v% aq. soln.) clear
Melting point
157°C to 161°C
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text