MEM
MEM
Gibco™

MEM

MEM (Minimum Essential Medium) is one of the most commonly used of all cell culture media. MEM can be usedMás información
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidad
110950721000 mL
11095080500 mL
1109509810 x 500 mL
110951146 x 1000 mL
Número de catálogo 11095072
Precio (MXN)
1,480.91
Each
Añadir al carro de la compra
Cantidad:
1000 mL
Customize this product
Precio (MXN)
1,480.91
Each
Añadir al carro de la compra
Ask our AI about this Product

MEM (Minimum Essential Medium) is one of the most commonly used of all cell culture media. MEM can be used with a variety of suspension and adherent mammalian cells, including HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, fibroblasts, and primary rat astrocytes. We offer a variety of MEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.


This MEM is modified as follows:
WithWithout
• L-glutamine• HEPES
• Phenol Red 


The complete formulation is available.

Using MEM
MEM was developed by Harry Eagle, based on his earlier formulation of Basal Medium Eagle (BME). Many other modifications of MEM followed, including Glasgow’s MEM, MEM α, DMEM, and Temin’s Modification. MEM is available with Earle’s salts for use in a CO2 incubator, or with Hanks' salts for use without CO2. This product is made with Earle’s salts. MEM contains no proteins, lipids, or growth factors. Therefore, MEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). MEM uses a sodium bicarbonate buffer system (2.2 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.

Para su uso en investigación o procesos de fabricación posteriores. No apto para uso diagnóstico ni para la administración directa en seres humanos ni en animales.

Especificaciones
Línea de célulasHeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, and fibroblasts
Tipo de célulaPrimary Rat Astrocytes
Concentración1 X
Calidad de fabricacióncGMP-compliant under the ISO 13485 standard
Línea de productosGibco™
Tipo de productoMEM (Minimum Essential Medium)
Cantidad1000 mL
Duración de almacenamiento12 Months From Date of Manufacture
Condiciones de envíoTemperatura ambiente
ClasificaciónAnimal Origin-free
FormularioLíquido
Serum LevelSuplementos de suero estándar
EsterilidadEstéril con filtro
Sterilization MethodSterile-filtered
Con aditivosLow Glucose, Glutamine, Phenol Red
Sin aditivosNo HEPES, No Sodium Pyruvate
Unit SizeEach
Contenido y almacenamiento
Condiciones de almacenamiento: De 2 a 8 °C. Proteger de la luz
Condiciones de envío: Ambiente
Vida útil: 12 meses a partir de la fecha de fabricación
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documentos y descargas

Certificados

Hojas de datos de seguridad

Compartir número de catálogo, nombre o enlace.