Thermo Scientific Chemicals

Cycloheptane, 99%, Thermo Scientific Chemicals

Catalog number: 111010050
5 mL, Glass bottle, Each
Thermo Scientific Chemicals

Cycloheptane, 99%, Thermo Scientific Chemicals

Catalog number: 111010050
5 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 111010050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
291-64-5
IUPAC Name
cycloheptane
Molecular Formula
C7H14
InChI Key
DMEGYFMYUHOHGS-UHFFFAOYSA-N
SMILES
C1CCCCCC1
GC
>=98.5 %
Appearance (Form)
Liquid
Refractive index
1.4440 to 1.4460 (20°C, 589 nm)
Appearance (Color)
Clear colorless
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text