Thermo Scientific Chemicals

Cycloheptanone, 98+%, Thermo Scientific Chemicals

Catalog number: 111045000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Cycloheptanone, 98+%, Thermo Scientific Chemicals

Catalog number: 111045000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 111045000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
502-42-1
IUPAC Name
cycloheptanone
Molecular Formula
C7H12O
InChI Key
CGZZMOTZOONQIA-UHFFFAOYSA-N
SMILES
O=C1CCCCCC1
Appearance (Color)
Clear colorless to yellow
Refractive index
1.4595 to 1.4635 (20°C, 589 nm)
Appearance (Form)
Liquid
GC
>=98.0 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text