Cycloheptylamine, 99%, Thermo Scientific Chemicals
Cycloheptylamine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cycloheptylamine, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number 111071000
Price (USD)/ Each
230.08
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
230.08
Add to cart
Cycloheptylamine, 99%, Thermo Scientific Chemicals
Catalog number111071000
Price (USD)/ Each
230.08
-
Add to cart
Chemical Identifiers
IUPAC Namecycloheptanamine
Molecular FormulaC7H15N
InChI KeyVXVVUHQULXCUPF-UHFFFAOYSA-N
SMILESNC1CCCCCC1
Molecular Weight (g/mol)113.20
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=98.5 %
Refractive index1.4710 to 1.4730 (20°C, 589 nm)
Water=<0.5 % (K.F.)
Appearance (Color)Clear colorless to light yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only